Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Antibody (16-10A1), Alexa Fluor™ 532, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143385AF532
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
B7-1/CD80 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD80 | |
The immunogen for this antibody was CD80. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
16-10A1 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Armenian Hamster | |
Protein A or G purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
941 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction