Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-1/CD80 Antibody (16-10A1) - BSA Free, Novus Biologicals™
SDP

Armenian Hamster Monoclonal Antibody

$340.50 - $682.00

Specifications

Antigen B7-1/CD80
Clone 16-10A1
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14338505
SDP
View Documents
Novus Biologicals
NBP1433850.05MG
0.05 mg
Each for $340.50
Only null left
Add to Cart
 
NBP14338501
SDP
View Documents
Novus Biologicals
NBP1433850.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
NBP1433855
SDP
View Documents
Novus Biologicals
NBP1433850.5MG
0.5 mg
Each for $682.00
Only null left
Add to Cart
 
Description

Description

B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

B7-1/CD80
0.5 mg/mL
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay
Unconjugated
RUO
Mouse
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
CD80
IgG
Affinity Purified
16-10A1
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional
Monoclonal
Armenian Hamster
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
941
The immunogen for this antibody was CD80.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.