Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Antibody (16C10), Alexa Fluor™ 647, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP312026AF647
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISASpecifications
B7-1/CD80 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG1 λ |
ELISA | |
16C10 | |
ELISA | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
This antibody was generated by immunization of cynomolgus monkeys with human B7.1 antigen. | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
941 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction