Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Hamster anti-Mouse, Clone: AP-MAB0713, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$206.50 - $524.00
Specifications
Antigen | B7-1/CD80 |
---|---|
Clone | AP-MAB0713 |
Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:100-300, Immunohistochemistry-Paraffin 1:100-300, CyTOF-ready |
Applications | Immunohistochemistry, Flow Cytometry, CyTOF, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019084
![]() |
Novus Biologicals
NBP2121780.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NBP2121781
![]() |
Novus Biologicals
NBP2121780.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
B7-1/CD80 | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:100-300, Immunohistochemistry-Paraffin 1:100-300, CyTOF-ready | |
Monoclonal | |
Purified | |
Mouse | |
Q00609 | |
941 | |
Mouse CD80 | |
Primary | |
Reconstitute with 0.4 ml sterilized PBS to a final concentration of 0.5 mg/ml. |
AP-MAB0713 | |
Immunohistochemistry, Flow Cytometry, CyTOF, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Hamster | |
A 0.2 um filtered solution in PBS with No Preservative | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
CD80 | |
IgG | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title