Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Antibody (IDEC-114 (Galiximab)), Biotin, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP281099B
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
B7-1/CD80 | |
Monoclonal | |
Biotin | |
PBS | |
CD80 | |
Galiximab was prepared by immunizing cynomolgus monkeys with recombinant B7-1/CD80 antigen. The variable regions of the light and heavy chains were then cloned by being incorporated into a cassette vector (N5LG1) containing human constant region genes and subsequently transfected into the Dg44 CHO cell line (Hariharan K, 2013) | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
941 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
IDEC-114 (Galiximab) | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Galiximab is a primatized mAb which consists of human constant and primate (cynomolgus macaque) variable regions and binds specifically to B7-1/CD80. B7-1/CD80 is a surface glycoprotein and a member of the B7 family of costimulatory molecules. B7-1/CD80 antigen regulates T cell activation (through interacting with CD28 or CD152) and is expressed transiently in antigen-presenting cells, T cells and normal B cells, and expressed consitutively on various subtypes of B-cell lymphomas. | |
Store at 4°C in the dark. | |
IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction