Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-1/CD80 Human anti-Human, Clone: 16C10, Novus Biologicals™

Human Monoclonal Antibody
$235.84 - $615.90
Specifications
| Antigen | B7-1/CD80 |
|---|---|
| Clone | 16C10 |
| Dilution | ELISA, Block/Neutralize |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for ELISA, NeutralizationSpecifications
| B7-1/CD80 | |
| ELISA, Block/Neutralize | |
| Unconjugated | |
| RUO | |
| Human | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| This antibody was generated by immunization of cynomolgus monkeys with human B7.1 antigen. | |
| Primary |
| 16C10 | |
| Monoclonal | |
| Human | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Signal Transduction | |
| PBS | |
| 941 | |
| IgG1 λ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title