Learn More
Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4) containing 10% glycerol. containingno Preservative |
| Gene ID (Entrez) | 941 |
| Name | Recombinant B7-1/CD80 Protein |
| Purification Method | >95%, by SDS-PAGE |
| Quantity | 20 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Endotoxin Concentration | < 1.0 EU per 1ug of protein (determined by LAL method) |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
