Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-2/CD86 Antibody (021), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290957V
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Cynomolgus Monkey samples. It is validated for ELISA.Specifications
B7-2/CD86 | |
Monoclonal | |
DyLight 405 | |
Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus B7-2/CD86 (Q9BDM4; Met1-His239). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology, Myeloid derived Suppressor Cell | |
942 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
21 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction