Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-2/CD86 Antibody (103), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289607G
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
B7-2/CD86 | |
Monoclonal | |
DyLight 488 | |
Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human B7-2/CD86 (NP_008820.2; Met1-His239). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology, Myeloid derived Suppressor Cell | |
942 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
103 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction