Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-2/CD86 Antibody (AP-MAB0803) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019087 0.025 mg
NB212182100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019087 Supplier Novus Biologicals Supplier No. NBP2121820.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 2 publications

B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen B7-2/CD86
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AP-MAB0803
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100-1000, Immunohistochemistry 1:-300-2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:-300-2000, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Gene Symbols CD86
Host Species Rat
Immunogen Mouse CD86
Purification Method Protein G purified
Quantity 0.025 mg
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.