Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse B7-2/CD86 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB741G100UG
This item is not returnable.
View return policy
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Mouse samples. It is validated for Flow Cytometry.Specifications
B7-2/CD86 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD86 | |
LPS-activated mouse B cells | |
100 μg | |
942 | |
Mouse | |
Supernatant |
Flow Cytometry | |
GL1 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse B7-1, recombinant human (rh) B7-1 or rhB7-2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction