Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H2/ICOSLG Antibody (599841) [Alexa Fluor™ 532], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB158X
Description
B7-H2/ICOSLG Monoclonal antibody specifically detects B7-H2/ICOSLG in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
B7-H2/ICOSLG | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
599841 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H2 , Glu47-Lys279, Accession # Q9JHJ8 | |
0.1 mL | |
Cell Cycle and Replication | |
23308 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction