Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H2/ICOSLG Antibody (ICOSL/3260), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279932AF488
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
B7-H2/ICOSLG | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ICOSLG | |
Recombinant fragment of human B7-H2/ICOSLG protein (around aa 23-149) (exact sequence is proprietary) (Uniprot: O75144) | |
0.1 mL | |
Cell Cycle and Replication | |
This monoclonal antibody recognizes a protein of 36kDa, identified as B7-H2/ICOSLG. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ICOSL/3260 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction