Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H2/ICOSLG Antibody (OTI2C5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271735H
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
B7-H2/ICOSLG | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ICOSLG | |
Full length recombinant protein of human B7-H2/ICOSLG (NP_056074) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2C5 | |
Western Blot, Immunohistochemistry | |
B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
23308.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction