Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3/CD276 Antibody (317), Alexa Fluor™ 532

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290756AF532
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for ELISA.Specifications
B7-H3/CD276 | |
Monoclonal | |
Alexa Fluor 532 | |
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse B7-H3/CD276 (NP_598744.1; Met1-Phe244). | |
0.1 mL | |
Immunology | |
80381 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
317 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction