Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3/CD276 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232250C
Description
B7-H3/CD276 Polyclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Western BlotSpecifications
B7-H3/CD276 | |
Polyclonal | |
Western Blot | |
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human CD276 Molecule using the numbering given in entry NP_001019907.1 (GeneID 80381). | |
0.1 mL | |
Immunology | |
80381 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction