Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3/CD276 Antibody (MM0104-20J12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212048F
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
B7-H3/CD276 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
MM0104-20J12 | |
Western Blot, Immunocytochemistry | |
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Human recombinant B7-H3/CD276 (extracellular domain) | |
0.1 mL | |
Immunology | |
80381 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction