Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3 (CD276) Mouse anti-Human, Unconjugated, Clone: 376.96, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1731100UL
Description
B7-H3 (CD276) Monoclonal antibody specifically detects B7-H3 (CD276) in Human samples. It is validated for Flow Cytometry, Inhibits Activity/Function, Immunoprecipitation, Western Blotting.
Specifications
B7-H3 (CD276) | |
Monoclonal | |
Unconjugated | |
CD276;4Ig-B7-H3;B7 homolog 3;B7-H3;Costimulatory molecule | |
Mouse | |
57.23 kDa calculated. | |
100 μL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunoprecipitation, Western Blot | |
376.96 | |
NP_079516 | |
CD276, B7H3, PSEC0249, UNQ309/PRO352 | |
Cultured COLO 38 human melanoma cells. | |
Protein G Purified | |
Inflammation and Immunology | |
Clone 376.96 is a mouse monoclonal antibody that specifically detects human B7-H3 (CD276). It targets an epitope within the extracellular domain. | |
Stable for 1 year at -20°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction