Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3 (CD276) Mouse anti-Human, Unconjugated, Clone: 376.96, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC173125UL
Description
B7-H3 (CD276) Monoclonal antibody specifically detects B7-H3 (CD276) in Human samples. It is validated for Flow Cytometry, Inhibits Activity/Function, Immunoprecipitation, Western Blotting.
Specifications
| B7-H3 (CD276) | |
| Monoclonal | |
| Unconjugated | |
| CD276;4Ig-B7-H3;B7 homolog 3;B7-H3;Costimulatory molecule | |
| Mouse | |
| 57.23 kDa calculated. | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| 376.96 | |
| NP_079516 | |
| CD276, B7H3, PSEC0249, UNQ309/PRO352 | |
| Cultured COLO 38 human melanoma cells. | |
| Protein G Purified | |
| Inflammation and Immunology | |
| Clone 376.96 is a mouse monoclonal antibody that specifically detects human B7-H3 (CD276). It targets an epitope within the extracellular domain. | |
| Stable for 1 year at -20°C from date of receipt. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction