Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H3 (CD276) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114525
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114525 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114525 Supplier Thermo Scientific Supplier No. PA5114525
Add to Cart
Add to Cart

B7-H3 (CD276) Polyclonal Antibody

B7-H3 (CD276) Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H3 (CD276)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CD276
Gene Accession No. Q5ZPR3, Q7TPB4, Q8VE98
Gene Alias 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352
Gene Symbols CD276
Host Species Rabbit
Immunogen CD276 antibody was raised against a peptide corresponding to 19 amino acids near the carboxy terminus of CD276. The immunogen is located within amino acids 484 - 534 of human CD276.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102657, 315716, 80381
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.