Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3/CD276 Rabbit anti-Mouse, Clone: 317, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | B7-H3/CD276 |
---|---|
Clone | 317 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for ELISA.Specifications
B7-H3/CD276 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
80381 | |
IgG | |
Protein A purified |
317 | |
ELISA | |
Unconjugated | |
Rabbit | |
Mouse | |
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Recombinant Mouse B7-H3/CD276 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title