Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H3/CD276 Antibody (MJ18) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2751950.025MG
Description
B7-H3/CD276 Monoclonal specifically detects B7-H3/CD276 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.Specifications
B7-H3/CD276 | |
Monoclonal | |
Unconjugated | |
PBS | |
CD276 | |
This antibody was generated by immunizing Sprague Dawley rats with B7-H3-Ig, consisting of the extracellular domain (aa1-242 of mouse B7-H3) linked to the Fc portion of mouse IgG2a, emulsified in CFA (Difco). The antibody was selected by its reactivity to mouse B7-H3-transfected cells, but not to parental cells by flow cytometry. | |
80381 | |
Mouse | |
Purified |
Flow Cytometry, ELISA, Blocking Assay, Neutralization | |
MJ18 | |
Flow Cytometry 1:100 - 1:1000, ELISA, Block/Neutralize | |
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
Rabbit | |
0.025 mg | |
This antibody binds to mouse B7-H3 and does not react with other B7-family molecules. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction