Learn More
B7-H3/CD276 Recombinant Rabbit Monoclonal Antibody (BLR026F), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $595.98
Specifications
Antigen | B7-H3/CD276 |
---|---|
Clone | BLR026F |
Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2646
![]() |
Bethyl Laboratories, Inc
A700026T |
10 μL |
Each for $157.08
|
|
|||||
50-157-2645
![]() |
Bethyl Laboratories, Inc
A700026 |
100 μL |
Each for $595.98
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommende
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.Specifications
B7-H3/CD276 | |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5ZPR3 | |
80381 | |
recombit protein representing the extracellular domain of human CD276 Molecule | |
Primary | |
4° C | |
CD276 |
BLR026F | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
PSEC0249; 4Ig-B7-H3; B7-H3; B7H3; B7RP-2; B7 homolog 3; CD276 antigen; costimulatory molecule; HB7H3 | |
CD276 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.