Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H4 Antibody (2582H), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB21542L
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal antibody specifically detects B7-H4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| B7-H4 | |
| Monoclonal | |
| DyLight 550 | |
| B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-H4, Phe29-Pro258, Accession # Q7TSP5 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 79679 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 2582H | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction