Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H4 Antibody (297219), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2154Z
Description
B7-H4 Monoclonal antibody specifically detects B7-H4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
B7-H4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
297219 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H4, Phe29-Pro258, Accession # Q7TSP5 | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
79679 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction