Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7-H4 Mouse anti-Human, Brilliant Violet 711, Clone: MIH43, BD Optibuild™

Catalog No. BDB742959 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742959 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742959 Supplier BD Biosciences Supplier No. 742959
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH43 monoclonal antibody specifically binds to human B7-H4 (B7 family member, H4) that is encoded by the VTCN1 (V-set domain containing T cell activation inhibitor 1) gene. B7-H4 is a type I membrane glycoprotein with a calculated molecular weight of 30.89 kDa. It is also known as VCTN1, B7h.5, B7S1 (B7 superfamily member 1) or B7X. B7-H4 is a newly discovered member of the B7 family of costimulatory proteins. B7-H4 is not constitutively expressed on peripheral tissues. Its expression can be induced on macrophages, dendritic cells, B cells and T cells. By binding to its putative receptor, the function of B7-H4 was initially reported to be a negative regulator of T cell activation, proliferation and differentiation related to cytokine production and cytotoxic effector cell functions. B7-H4 is reportedly overexpressed in a variety tumors. B7-H4 expression by tumor macrophages appears to play a role in suppression antigen-specific T cell mediated immunity. Recent studies indicate that B7-H4 can also be a positive regulator of T cell responses. B7-H4 can be involved innate immunity as well, eg, by inhibiting neutrophil expansion.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H4
Classification Monoclonal
Clone MIH43
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias VTCN1, VCTN1, B7 family member, H4, B7H4, B7h.5, B7S1, B7X
Host Species Mouse
Immunogen Recombinant Human B7-H4 protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.