Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H6 Antibody (1002932), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10127MFV500
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOFSpecifications
| B7-H6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Immunology | |
| 374383 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 1002932 | |
| Flow Cytometry, CyTOF-ready | |
| B7 homolog 6, B7-H6, DKFZp686O24166 | |
| Human embryonic kidney cell, HEK293-derived cynomolgus Monkey B7-H6, Asp25-Ser262 | |
| RUO | |
| Primary | |
| Cynomolgus Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction