Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H6 Antibody (2167B), PE/Atto594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB71443PEATT594
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow CytometrySpecifications
B7-H6 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2167B | |
Flow Cytometry | |
B7 homolog 6, B7-H6, DKFZp686O24166 | |
Human embryonic kidney cell, HEK293-derived human B7-H6 protein, Asp25-Ser262, Accession # Q68D85 | |
0.1 mL | |
Cancer, Immunology | |
374383 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction