Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H6 Antibody (875023), PE/Atto594, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB71441PEATT594
Description
B7-H6 Monoclonal specifically detects B7-H6 in Human samples. It is validated for Flow Cytometry.Specifications
B7-H6 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
NCR3LG1 | |
Partial recombinant human B7-H6 protein (amino acids1-262). [UniProt Q68D85] | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
875023 | |
Flow Cytometry | |
B7 homolog 6, B7-H6, DKFZp686O24166 | |
Mouse | |
Protein A or G purified | |
Cancer, Immunology | |
374383.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction