Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B7-H6 Chimeric Recombinant Rabbit Monoclonal Antibody (17B1.3)
GREENER_CHOICE

Catalog No. PIMA548287
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548287 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548287 Supplier Invitrogen™ Supplier No. MA548287
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: mAb 17B1.3 specifically recognises B7-H6. 17B1.3 reacted with human tumour cell lines constitutively expressing B7-H6 such as HeLa cells. Fab 17B1.3 does not interfere with the binding of B7-H6 to NKp30, but nevertheless blocks NK cell activation.

B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H6 Chimeric
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 17B1.3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene NCR3LG1
Gene Accession No. Q68D85
Gene Alias B7 homolog 6; B7H6; B7-H6; DKFZp686I21167; DKFZp686O24166; natural cytotoxicity triggering receptor 3 ligand 1; natural killer cell cytotoxicity receptor 3 ligand 1; NCR3LG1; putative Ig-like domain-containing protein DKFZp686O24166/DKFZp686I21167
Gene Symbols NCR3LG1
Host Species Rabbit
Immunogen Immunised mice with extracellular domain of human B7-H6 and selected on the basis of specific reactivity by P815.B7-H6, but not with P815.B7-H1.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374383
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.