Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H6 Monoclonal Antibody (JAM1EW), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$514.00
Specifications
Antigen | B7-H6 |
---|---|
Clone | JAM1EW |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
46-652-642
![]() |
Invitrogen™
46652642 |
100 Tests |
Each of 1 for $514.00
|
![]() |
|||||
Description
Mouse Monoclonal Antibody
B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.Specifications
B7-H6 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q68D85 | |
NCR3LG1 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
JAM1EW | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
374383 | |
B7-H6 recombinant protein. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
NCR3LG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title