Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H6 Rat anti-Human, BV421, Clone: 1A5, BD Horizon™
Rat Monoclonal Antibody
$215.00 - $488.00
Specifications
Antigen | B7-H6 |
---|---|
Clone | 1A5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
B7-H6 Monoclonal antibody specifically detects B7-H6 in Human samples. It is validated for Flow cytometrySpecifications
B7-H6 | |
Flow Cytometry | |
Brilliant Violet 421 | |
Rat | |
Human | |
B7 homolog 6; B7-H6; B7H6; DKFZp686O24166; NCR3LG1 | |
Human B7-H6 Transfected Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
1A5 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
374383 | |
IgM κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title