Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H7/HHLA2 Antibody (907812R) [Biotin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB80841RB
Description
B7-H7/HHLA2 Monoclonal antibody specifically detects B7-H7/HHLA2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
B7-H7/HHLA2 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
907812R | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2, Met1-Asn344, Accession # Q9UM44 | |
0.1 mL | |
Immunology, Inflammation | |
11148 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction