Learn More
B7-H7 Monoclonal Antibody (MA57YW), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$425.00
Specifications
Antigen | B7-H7 |
---|---|
Clone | MA57YW |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-653-742
![]() |
Invitrogen™
17653742 |
100 Tests |
Each of 1 for $425.00
|
![]() |
|||||
Description
Description: This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells. Applications Reported: This MA57YW antibody has been reported for use in flow cytometric analysis. Applications Tested: This MA57YW antibody has been pre-diluted and tested by flow cytometric analysis of the HDLM2 cell line. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.Specifications
B7-H7 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9UM44 | |
HHLA2 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MA57YW | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
11148 | |
B7-H7 recombinant protein. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
HHLA2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.