Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B7-H7 Monoclonal Antibody (MA57YW), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$497.00
Specifications
| Antigen | B7-H7 |
|---|---|
| Clone | MA57YW |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
61-653-742
![]()
|
Invitrogen™
61653742 |
100 Tests |
Each of 1 for $497.00
|
|
|||||
Description
Mouse Monoclonal Antibody
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.Specifications
| B7-H7 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q9UM44 | |
| HHLA2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MA57YW | |
| Flow Cytometry | |
| PE-eFluor 610 | |
| Mouse | |
| Human | |
| 11148 | |
| B7-H7 recombinant protein. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| HHLA2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title