Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B7H4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980436
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0436 Supplier Bioss Supplier No. BS0673R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B7-H4 (B7x, VTCN1) a costimulatory protein which is reported to function as a negative regulator of T-cell mediated immunity. Although B7-H4 binds an unknown receptor, it is thought to deliver an inhibitory signal to T-cells preventing their proliferation, cell cycle progression and interleukin-2 production. B7-H4 deficient mice are only minimally affected; suggesting B7-H4 is important in the fine tuning of the T-cell mediated immune response. B7-H4 is expressed on activated T-cells, B-cells, monocytes and dendritic cells. Aberrant expression has been associated with cancers of the lung, breast and ovary in humans. B7-H4 is expressed primarily on the membrane of lymphoid cells, and as an immunoinhibitory protein that interacts with receptors on the surface of T lymphocytes, it is involved in mediating cellular and humoral immune responses. Overexpression of B7-H4 is associated with certain malignancies, including ovarian and breast cancer, which may be a mechanism by which tumor cells suppress T cell immunity and facilitate tumor progression. Further, VTCN1 may be a useful biomarker for the early detection of ovarian cancer and Renal Cell Carcinoma (RCC).

Specifications

Antigen B7H4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Vtcn1
Gene Accession No. Q7Z7D3
Gene Alias B7 family member, H4; B7 homolog 4; B7 superfamily member 1; B7h.5; B7H4; B7-H4; B7s1; B7X; BC032925; EMBL:AAH95842.1}; FLJ22418; Immune costimulatory protein B7-H4; PRO1291; Protein B7S1; RGD1311204; RP11-229A19.4; T cell costimulatory molecule B7x; T-cell costimulatory molecule B7x; UNQ659/PRO1291; VCTN1; V-set domain containing T cell activation inhibitor 1; V-set domain containing T-cell activation inhibitor 1; V-set domain-containing T-cell activation inhibitor 1; VTCN1; vtcn1 {ECO:0000312
Gene Symbols Vtcn1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Rat B7H4 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79679
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.