Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B9D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554645 Supplier Invitrogen™ Supplier No. PA554645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPSVFLLMV NGQVESAQFP EYDDLYCKYC FVYGQDWAPT AGLEEGISQI TSKSQDVRQA LVWN Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 98%.

This gene encodes a B9 domain-containing protein, one of several that are involved in ciliogenesis. Alterations in expression of this gene have been found in a family with Meckel syndrome. Meckel syndrome has been associated with at least six different genes. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Three alternatively spliced transcript variants that encode different proteins have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B9D1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene B9D1
Gene Accession No. Q9UPM9
Gene Alias AW045994; B9; B9 domain containing 1; B9 domain-containing protein 1; B9 protein domain 1; B9D1; Endothelial precursor cells protein B9; endothelial precursor protein B9; Eppb9; MKS1-related protein 1; MKS9; MKSR1
Gene Symbols B9D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human B9D1. Recombinant protein control fragment (Product #RP-93202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.