Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BA Antigen (Leu21) Antibody (DF1588), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254564PEATT594
Description
BA Antigen (Leu21) Monoclonal specifically detects BA Antigen (Leu21) in Human samples. It is validated for Flow Cytometry.Specifications
BA Antigen (Leu21) | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
DF1588 | |
Flow Cytometry | |
CD78, CD78, Cdw78, Cdw78, LO panB a | |
Human leucocytes | |
0.1 mL | |
Reacts with a carbohydrate determinant, identified as CDw78 (Workshop V; Code CDw78.2). Epitope recognized by this monoclonal antibody and Leu21 is the same. It is present on some immature and some peripheral blood B cells, and some B cell lines. Its expression is increased after B-cell activation in vitro. CDw78 is also found on tissue macrophages and on epithelial cells, but not on T cells, NK cells, monocytes, granulocytes and myeloid tissues. It appears that CDw78 is a determinant on a major histocompatibility complex (MHC) class II subpopulation. | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction