Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572197
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572197 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572197 Supplier Invitrogen™ Supplier No. PA572197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BA1 Polyclonal Antibody for Western Blot

Involved in oxygen transport from gills to the various peripheral tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ba1
Gene Accession No. Q90486
Gene Alias ba1; bA1 globin; ba1l; ba1l protein; ba2; ba2 globin; ba2 globin, like; ba2l; Beta-1-globin; beta-2-globin; beta-A1-globin; beta-A2-globin; hbb; hbba1; hbba1.2; Hemoglobin beta-1 chain; hemoglobin beta-2 chain; hemoglobin subunit beta-1; hemoglobin subunit beta-2; hemoglobin, beta adult 1; si:bY187G17.2; si:by187g17.4; si:dZ118J2.3; zgc:123164; zgc:136512
Gene Symbols ba1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 2-35 amino acids from the N-terminal region of DANRE ba1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30216
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.