Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAALC Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1002413
Description
BAALC Polyclonal antibody specifically detects BAALC in Mouse samples. It is validated for Western Blot, ELISA.Specifications
BAALC | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
brain and acute leukemia cytoplasmic protein, brain and acute leukemia, cytoplasmic, FLJ12015 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_079088.1) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.03 - 0.1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
NP_079088.1 | |
BAALC | |
Peptide with sequence C-KQNGLQTTEAKRDAK corresponding to internal region according to NP_079088.1. | |
0.1 mg | |
Neuroscience | |
79870 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction