Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAALC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518435
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518435 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518435 Supplier Invitrogen™ Supplier No. PA518435
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This gene was identified by gene expression studies in patients with acute myeloid leukemia. The gene is conserved among mammals and is not found in lower organisms. Tissues that express this gene develop from the neuroectoderm. Multiple alternatively spliced transcript variants that encode different proteins have been described for this gene; however, some of the transcript variants are found only in AML cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAALC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BAALC
Gene Accession No. Q8VHV1, Q8WXS3
Gene Alias 2810457D07Rik; Baalc; Brain and acute leukemia cytoplasmic protein; brain and acute leukemia, cytoplasmic; FLJ12015; HGNC:14333
Gene Symbols BAALC
Host Species Goat
Immunogen Synthetic peptide sequence (KQNGLQTTEAKRDAK) corresponding to the internal amino acids of BAALC (aa 101-115).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118452, 79870
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.