Learn More
BAALC Rabbit anti-Human,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | BAALC |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | KLH |
| Gene Alias | Brain and acute leukemia cytoplasmic protein |
| Gene Symbols | BAALC |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Brain and acute leukemia cytoplasmic protein (BAALC). |
| Molecular Weight of Antigen | ∼20 kDa observed; 19.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.