Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAALC Rabbit anti-Human,Mouse,Rat, KHL, Polyclonal, MilliporeSigma™

Catalog No. ABN224325MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN224325MI 25 μg
ABN224310MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN224325MI Supplier MilliporeSigma Supplier No. ABN224325UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAALC Polyclonal antibody specifically detects BAALC in Human,Mouse,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAALC
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate KLH
Gene Alias Brain and acute leukemia cytoplasmic protein
Gene Symbols BAALC
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Brain and acute leukemia cytoplasmic protein (BAALC).
Molecular Weight of Antigen ∼20 kDa observed; 19.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody that detects Brain and acute leukemia cytoplasmic protein (BAALC). It targets an epitope within 16 amino acids from the N-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.