Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAALC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989394
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9394 Supplier Bioss Supplier No. BS9849R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene was identified by gene expression studies in patients with acute myeloid leukemia. The gene is conserved among mammals and is not found in lower organisms. Tissues that express this gene develop from the neuroectoderm. Multiple alternatively spliced transcript variants that encode different proteins have been described for this gene; however, some of the transcript variants are found only in AML cell lines.

Specifications

Antigen BAALC
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BAALC
Gene Accession No. Q8WXS3
Gene Alias 2810457D07Rik; Baalc; Brain and acute leukemia cytoplasmic protein; brain and acute leukemia, cytoplasmic; FLJ12015; HGNC:14333
Gene Symbols BAALC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BAALC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79870
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.