Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529060
Description
Recommended positive controls: A431, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a liver enzyme that catalyzes the transfer of C24 bile acids from the acyl-CoA thioester to either glycine or taurine, the second step in the formation of bile acid-amino acid conjugates. The bile acid conjugates then act as a detergent in the gastrointestinal tract, which enhances lipid and fat-soluble vitamin absorption. Defects in this gene are a cause of familial hypercholanemia (FHCA). Two transcript variants encoding the same protein have been found for this gene.
Specifications
| BAAT | |
| Polyclonal | |
| Unconjugated | |
| BAAT | |
| AI118337; AI158864; Baat; BACAT; BAT; bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase); bile acid CoA:amino acid N-acyltransferase; bile acid Coenzyme A: amino acid N-acyltransferase (glycine N-choloyltransferase); Bile acid-CoA thioesterase; Bile acid-CoA:amino acid N-acyltransferase; bile acid-Coenzyme A dehydrogenase: amino acid n-acyltransferase; bile acid-Coenzyme A: amino acid N-acyltransferase; Choloyl-CoA hydrolase; Glycine N-choloyltransferase; kan-1; Long-chain fatty-acyl-CoA hydrolase; taurine N-acyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 570 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14032 | |
| BAAT | |
| Recombinant protein encompassing a sequence within the center region of human BAAT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction