Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596744 Supplier Invitrogen™ Supplier No. PA596744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YLFPIEEAQG QFLFIVGEGD KTINSKAHAE QAIGQLKRHG KNNWTLLSYP GAGHLIEPPY SPLCCASTTH DLRLHWGGEV IPHAAAQEHA WKEIQRFLRK HLIPDVTSQL; Positive Samples: Jurkat, BT-474, A-549, HepG2, Mouse liver, Rat liver; Cellular Location: Cytoplasm.

The protein encoded by this gene is a liver enzyme that catalyzes the transfer of C24 bile acids from the acyl-CoA thioester to either glycine or taurine, the second step in the formation of bile acid-amino acid conjugates. The bile acid conjugates then act as a detergent in the gastrointestinal tract, which enhances lipid and fat-soluble vitamin absorption. Defects in this gene are a cause of familial hypercholanemia (FHCA). Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAAT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BAAT
Gene Accession No. Q14032, Q63276, Q91X34
Gene Alias AI118337; AI158864; Baat; BACAT; BAT; bile acid CoA: amino acid N-acyltransferase (glycine N-choloyltransferase); bile acid CoA:amino acid N-acyltransferase; bile acid Coenzyme A: amino acid N-acyltransferase (glycine N-choloyltransferase); Bile acid-CoA thioesterase; Bile acid-CoA:amino acid N-acyltransferase; bile acid-Coenzyme A dehydrogenase: amino acid n-acyltransferase; bile acid-Coenzyme A: amino acid N-acyltransferase; Choloyl-CoA hydrolase; Glycine N-choloyltransferase; kan-1; Long-chain fatty-acyl-CoA hydrolase; taurine N-acyltransferase
Gene Symbols BAAT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 309-418 of human BAAT (NP_0016921).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12012, 29725, 570
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.