Learn More
BAAT1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | BAAT1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1496
![]() |
Bethyl Laboratories, Inc
IHC00104T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1495
![]() |
Bethyl Laboratories, Inc
IHC00104 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Crab-eating macaque
Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.Specifications
BAAT1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
BAAT1, C7orf27, MGC22916, BRCA1-associated protein required for ATM activation protein 1; HEAT repeat-containing protein C7orf27 | |
BRAT1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q6PJG6 | |
221927 | |
Between 775 and C-term | |
Primary | |
4° C | |
BRAT1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.