Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAAT1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558318
Encompass_Preferred
Change view
Click to view available options
20 μl; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-155-8318 20 μl; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8318 Supplier Bethyl Laboratories, Inc Supplier No. A300728AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Required for activation of ATM following ionizing radiation. May act by regulating dephosphorylation of ATM.

Specifications

Antigen BAAT1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene BRAT1
Gene Accession No. Q6PJG6
Gene Alias BAAT1, C7orf27, MGC22916, BRCA1-associated protein required for ATM activation protein 1; HEAT repeat-containing protein C7orf27
Gene Symbols BRAT1
Host Species Rabbit
Immunogen Between 775 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221927
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.