Learn More
Invitrogen™ BABAM1 Monoclonal Antibody (GT2810)

Description
Positive Control: 293T, A431, HeLa, HepG2 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | BABAM1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT2810 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | BABAM1 |
| Gene Accession No. | Q9NWV8 |
| Gene Alias | 5430437P03Rik; BABAM1; BRCA1-A complex subunit MERIT40; BRISC and BRCA1 A complex member 1; BRISC and BRCA1-A complex member 1; C19orf62; HSPC142; mediator of Rap80 interactions and targeting 40 kDa; mediator of RAP80 interactions and targeting subunit of 40 kDa; MERIT40; NBA1; new component of the BRCA1 A complex; new component of the BRCA1-A complex; new component of the BRCAA1 A complex |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.