Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Baboon Rabbit anti-Drosophila, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc BAB101AP

Description
Baboon Polyclonal antibody specifically detects Baboon in Drosophila samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
Baboon | |
Polyclonal | |
Unconjugated | |
babo | |
Rabbit | |
Affinity chromatography | |
RUO | |
35900 | |
-20°C | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
babo; CG8224; Receptor protein serine/threonine kinase | |
N-epitope Synthetic peptide corresponding to amino acid region 200-221 of Drosophila melanogaster protein. The peptide was covalently modified for conjugation.Synthetic peptide:CNKNENMKRIFEDYVPHRLTS | |
200 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction